Assay Details
| Gene Symbol: ZMIZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097263.1 | - | 1 | 1550 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | oxoglutarate dehydrogenase | OGDH |
AKGDH, E1k, OGDC |
-60.8kb | Sense | Chr. 7 - 44606522 - 44709070 on NCBI Build GRCh38 |
| Downstream | peptidylprolyl isomerase A | PPIA |
CYPA, CYPH, HEL-S-69p |
24.9kb | Sense | Chr. 7 - 44796636 - 44803123 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZMIZ2
Genomic Map