Assay Details
| Gene Symbol: SBF2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098060.1 | - | 1 | 1346 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC440028 | LOC440028 | Anti-sense | Chr. 11 - 9754770 - 9759533 on NCBI Build GRCh38 | ||
| Downstream | SET binding factor 2 | SBF2 |
CMT4B2, DENND7B, MTMR13 |
18.7kb | Anti-sense | Chr. 11 - 9778667 - 10294216 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SBF2-AS1
Genomic Map