| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015013.hg.1 |
ENST00000407505.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015013.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015013.hg.1 |
TC22000896.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015015.hg.1 |
ENST00000407505.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015015.hg.1 |
ENST00000467207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015015.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015015.hg.1 |
TC22000896.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015014.hg.1 |
ENST00000407505.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015014.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015014.hg.1 |
TC22000896.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015018.hg.1 |
ENST00000407505.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015018.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015018.hg.1 |
TC22000896.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000630584.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
ENST00000630584.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165622.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165622.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165622.hg.1 |
TC2200009078.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165622.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165622.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000630584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
ENST00000630584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009078.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165623.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165623.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165623.hg.1 |
ENST00000630584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165623.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165623.hg.1 |
TC2200009078.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165623.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165623.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165623.hg.1 |
ENST00000630584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165623.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165621.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165621.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165621.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165621.hg.1 |
TC2200009078.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165621.hg.1 |
ENST00000625694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165621.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165621.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165625.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165625.hg.1 |
NR_038917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165625.hg.1 |
TC2200009078.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165625.hg.1 |
ENST00000626321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165625.hg.1 |
NR_038917.1 |
Refseq |