Assay Details
| Gene Symbol: NOL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC037841.1 | - | 1 | 1647 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929715 | LOC101929715 | -215.9kb | Anti-sense | Chr. 2 - 10449728 - 10455552 on NCBI Build GRCh38 | |
| Downstream | ATPase H+ transporting V1 subunit C2 | ATP6V1C2 |
ATP6C2, VMA5 |
47.8kb | Anti-sense | Chr. 2 - 10720974 - 10785110 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NOL10
Genomic Map