Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 9 member A6 | SLC9A6 |
MRSA, NHE6 |
-110.1kb | Sense | Chr. X - 135974597 - 136047269 on NCBI Build GRCh38 |
| Downstream | MAP7 domain containing 3 | MAP7D3 | MDP3 | 54.1kb | Anti-sense | Chr. X - 136213220 - 136256482 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map