Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transient receptor potential cation channel subfamily V member 6 | TRPV6 |
ABP/ZF, CAT1, CATL, ECAC2, HSA277909, LP6728, ZFAB |
Sense | Chr. 7 - 142871203 - 142885745 on NCBI Build GRCh38 | |
| Downstream | transient receptor potential cation channel subfamily V member 5 | TRPV5 |
CAT2, ECAC1, OTRPC3 |
33.7kb | Sense | Chr. 7 - 142908101 - 142933723 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map