Assay Details
| Gene Symbol: NUDT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC042967.1 | - | 1 | 1448 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102467212 | LOC102467212 | -19.1kb | Sense | Chr. 5 - 103528434 - 103541985 on NCBI Build GRCh38 | |
| Downstream |
RAB9B, member RAS oncogene family pseudogene 1 |
RAB9BP1 | RAB9P1 | 1.5mb | Anti-sense | Chr. 5 - 105099474 - 105100098 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NUDT12
Genomic Map