Assay Details
| Gene Symbol: IPO11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC043163.2 | - | 1 | 1251 | 122 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DIM1 dimethyladenosine transferase 1 homolog | DIMT1 |
DIM1, DIMT1L, HSA9761, HUSSY5 |
-80.7kb | Sense | Chr. 5 - 62388523 - 62403939 on NCBI Build GRCh38 |
| Downstream | leucine rich repeat containing 70 | LRRC70 | SLRN | 92.8kb | Anti-sense | Chr. 5 - 62578735 - 62581448 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

IPO11
Genomic Map