Assay Details
| Gene Symbol: TCEANC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097032.1 | - | 2 - 3 | 968 | 77 | |
| BC044219.1 | - | 1 - 2 | 393 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream |
RAB9A, member RAS oncogene family |
RAB9A | RAB9 | 20.4kb | Sense | Chr. X - 13689121 - 13709825 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

TCEANC
Genomic Map