Assay Details
| Gene Symbol: LOC105378146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC044924.2 | - | 1 | 1449 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SPARC related modular calcium binding 2 | SMOC2 |
DTDP1, MST117, MSTP117, MSTP140, SMAP2, bA270C4A.1, bA37D8.1, dJ421D16.1 |
-294.6kb | Anti-sense | Chr. 6 - 168441151 - 168667994 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929460 | LOC101929460 | 69.8kb | Sense | Chr. 6 - 169034171 - 169035643 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460148 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252084 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460181 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252108 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252070 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460163 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460157 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252065 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252077 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252125 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460198 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460176 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460194 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252112 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460165 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460185 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460173 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460195 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252067 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252085 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460166 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460149 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460167 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460183 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252071 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460171 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252072 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252063 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252068 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460153 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252073 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460172 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460143 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460155 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460188 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460200 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460180 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252101 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460168 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252137 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460142 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252120 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252128 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460159 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460152 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252069 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460179 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460182 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460147 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460191 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460156 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252115 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252109 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460196 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460158 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25460186 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05252129 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 573462 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3576317 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24776 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3576317 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 244120_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013985.hg.1 | ENST00000444586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013985.hg.1 | ENST00000444586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013985.hg.1 | ENST00000444586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013985.hg.1 | ENST00000444586.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 244120_at | - | - |
| Human Genome U133 Plus 2.0 Array | 244120_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244120_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244120_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244120_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244120_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244120_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244120_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244120_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244120_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244120_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244120_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244120_at2 | - | - |
| Human Genome U133 Set | 244120_PM_at | - | - |
| Human Genome U133 Set | 244120_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina human-6 v2.0 expression beadchip | ILMN_1907883 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 1500392 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_86808 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1907883 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1907883 | - | - |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1907883 | - | - |

LOC105378146
Genomic Map