Assay Details
| Gene Symbol: ZDHHC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC050652.1 | - | 1 | 1354 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | exosome component 1 | EXOSC1 |
CGI-108, CSL4, Csl4p, SKI4, Ski4p, p13 |
-176.0 | Anti-sense | Chr. 10 - 97435909 - 97446065 on NCBI Build GRCh38 |
| Downstream |
MMS19 homolog, cytosolic iron-sulfur assembly component |
MMS19 |
MET18, MMS19L, hMMS19 |
10.4kb | Anti-sense | Chr. 10 - 97458324 - 97498794 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZDHHC16
Genomic Map