Assay Details
| Gene Symbol: LOC101929709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125822.1 | 5 | 1551 | 93 | ||
| GenBank mRNA | BQ007151.1 | - | 1 | 93 | ||
| BX538329.1 | - | 1 | 3751 | 93 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BU728465.1 | - | 1 | Chr. 8 - 89717399 - 89717962 [+] on Build GRCh38 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | matrix metallopeptidase 16 | MMP16 |
C8orf57, MMP-X2, MT-MMP2, MT-MMP3, MT3-MMP |
-1.4mb | Sense | Chr. 8 - 88037232 - 88327488 on NCBI Build GRCh38 |
| Downstream | receptor interacting serine/threonine kinase 2 | RIPK2 |
CARD3, CARDIAK, CCK, GIG30, RICK, RIP2 |
36.5kb | Anti-sense | Chr. 8 - 89757747 - 89791064 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3514859 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187625.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187625.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187625.hg.1 | ENST00000621883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800187625.hg.1 | NR_125822.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800187625.hg.1 | TC0800012438.hg.1 | Clariom D TC |

LOC101929709
Genomic Map