Assay Details
| Gene Symbol: SPINK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX648792.1 | - | 1 | 3053 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 5 open reading frame 46 | C5orf46 | SSSP1 | -180.0kb | Anti-sense | Chr. 5 - 147850463 - 147906594 on NCBI Build GRCh38 |
| Downstream |
serine peptidase inhibitor, Kazal type 14 (putative) |
SPINK14 | SPINK5L2 | 78.5kb | Sense | Chr. 5 - 148146372 - 148176076 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SPINK5
Genomic Map