Assay Details
| Gene Symbol: LOC100130865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123130.1 | - | 1 | 2854 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MAX dimerization protein 4 | MXD4 |
MAD4, MST149, MSTP149, bHLHc12 |
-1.0kb | Sense | Chr. 4 - 2247432 - 2262143 on NCBI Build GRCh38 |
| Downstream | zinc finger FYVE-type containing 28 | ZFYVE28 |
LST2, LYST2 |
1.0kb | Sense | Chr. 4 - 2269585 - 2418698 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100130865
Genomic Map