Assay Details
| Gene Symbol: LOC100128429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123323.1 | - | 1 | 1757 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 640 | MIR640 |
MIRN640, hsa-mir-640 |
-23.8kb | Anti-sense | Chr. 19 - 19435063 - 19435158 on NCBI Build GRCh38 |
| Downstream | testis specific serine kinase 6 | TSSK6 |
CT72, FKSG82, SSTK, TSSK4 |
53.3kb | Sense | Chr. 19 - 19514219 - 19515660 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100128429
Genomic Map