Assay Details
| Gene Symbol: LOC100130817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123329.1 | - | 1 | 3250 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NLR family CARD domain containing 5 | NLRC5 |
CLR16.1, NOD27, NOD4 |
-57.4kb | Anti-sense | Chr. 16 - 56989547 - 57083524 on NCBI Build GRCh38 |
| Downstream | family with sequence similarity 192 member A | FAM192A |
C16orf94, CDA018, CDA10, NIP30 |
7.9kb | Sense | Chr. 16 - 57152466 - 57186242 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100130817
Genomic Map