| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008678.hg.1 |
ENST00000547077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008678.hg.1 |
TC10000735.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000434878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000434878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000434878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000434878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000456391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000456391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000456391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000456391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000456391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000456391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000547077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000547077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
ENST00000547077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008676.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008676.hg.1 |
ENST00000547077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008676.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008676.hg.1 |
TC10000735.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000159317.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159317.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159317.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159317.hg.1 |
ENST00000547077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159317.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159317.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159317.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159317.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159317.hg.1 |
ENST00000547077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159317.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159317.hg.1 |
TC1000008678.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000430651.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000434878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000434878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000434878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000434878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000454527.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000456391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000456391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000456391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000456391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000456391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000456391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000547077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000547077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
ENST00000547077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008678.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159314.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159314.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159314.hg.1 |
ENST00000547077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159314.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159314.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159314.hg.1 |
ENST00000546988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159314.hg.1 |
ENST00000547077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159314.hg.1 |
NR_029380.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159314.hg.1 |
TC1000008678.hg.1 |
Clariom D TC |