Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin conjugating enzyme E2 T | UBE2T |
FANCT, HSPC150, PIG50 |
-208.7kb | Anti-sense | Chr. 1 - 202331657 - 202341966 on NCBI Build GRCh38 |
| Downstream | synaptotagmin 2 | SYT2 |
CMS7, MYSPC, SytII |
36.4kb | Anti-sense | Chr. 1 - 202590596 - 202710444 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map