Assay Details
| Gene Symbol: LOC100132481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130567.1 | - | 1 | 451 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil-helix-coiled-coil-helix domain containing 6 | CHCHD6 |
CHCM1, Mic25, PPP1R23 |
Sense | Chr. 3 - 126704220 - 126960420 on NCBI Build GRCh38 | |
| Downstream | plexin A1 | PLXNA1 |
NOV, NOVP, PLEXIN-A1, PLXN1 |
273.5kb | Sense | Chr. 3 - 126982119 - 127037392 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100132481
Genomic Map