Assay Details
| Gene Symbol: LOC100128482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130731.1 | - | 1 | 747 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 5003 | MIR5003 | -2.9kb | Sense | Chr. 5 - 172662165 - 172662263 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928093 | LOC101928093 | 88.2kb | Sense | Chr. 5 - 172755503 - 172762562 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648940 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469487 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648931 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648975 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469539 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469556 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648945 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648965 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469545 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469570 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648919 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469536 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648894 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648970 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648963 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469543 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469489 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469532 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469519 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469499 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469520 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469551 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469565 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469505 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648958 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648954 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469481 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469560 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648921 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648961 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469528 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648923 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648944 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648982 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469537 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469483 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469501 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469504 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469562 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469524 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469515 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469548 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648890 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648959 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469522 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15763863 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648953 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648948 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648896 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469507 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469503 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648937 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648980 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469484 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469486 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469482 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469553 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469561 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469557 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469517 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469566 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648904 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04469497 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24648951 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3216625 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 1441 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3216625 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3216625 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01396638 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 105067 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01396638 | - | - |

LOC100128482
Genomic Map