Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 16 member 3 | SLC16A3 |
MCT 3, MCT 4, MCT-3, MCT-4, MCT3, MCT4 |
-16.8kb | Sense | Chr. 17 - 82228406 - 82239499 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929511 | LOC101929511 | 31.8kb | Anti-sense | Chr. 17 - 82290046 - 82292814 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map