Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pleckstrin homology like domain family A member 3 | PHLDA3 | TIH1 | -35.9kb | Sense | Chr. 1 - 201464284 - 201469171 on NCBI Build GRCh38 |
| Downstream | ribosomal protein S10 pseudogene 7 | RPS10P7 | RPS10_2_147 | 13.5kb | Anti-sense | Chr. 1 - 201519904 - 201520602 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map