Assay Details
| Gene Symbol: THRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124042.1 | - | 1 | 1346 | 97 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DB359513.1 | - | 1 | Chr. 3 - 24115920 - 24116451 [+] on Build GRCh38 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 691 | LINC00691 | -12.7kb | Sense | Chr. 3 - 24099974 - 24103247 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101927854 | LOC101927854 | 33.7kb | Anti-sense | Chr. 3 - 24151324 - 24189956 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

THRB
Genomic Map