Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium sodium-activated channel subfamily T member 2 | KCNT2 |
KCa4.2, SLICK, SLO2.1 |
-93.8kb | Anti-sense | Chr. 1 - 196225779 - 196608905 on NCBI Build GRCh38 |
| Downstream | complement factor H related 3 | CFHR3 |
CFHL3, DOWN16, FHR-3, FHR3, HLF4 |
71.9kb | Sense | Chr. 1 - 196774800 - 196794073 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map