Assay Details
| Gene Symbol: PAOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124186.1 | - | 1 | 1657 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
enoyl-CoA hydratase, short chain 1 |
ECHS1 |
ECHS1D, SCEH |
-8.2kb | Anti-sense | Chr. 10 - 133362483 - 133373404 on NCBI Build GRCh38 |
| Downstream | mitochondrial ribosome associated GTPase 1 | MTG1 |
GTP, GTPBP7 |
10.5kb | Sense | Chr. 10 - 133394117 - 133420670 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PAOX
Genomic Map