Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105374344 | LOC105374344 | -31.6kb | Sense | Chr. 4 - 1113639 - 1132975 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100130872 | LOC100130872 | 27.6kb | Anti-sense | Chr. 4 - 1195783 - 1208962 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map