Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase) | LRAT | LCA14 | Sense | Chr. 4 - 154740841 - 154753119 on NCBI Build GRCh38 | |
| Downstream | RNA binding motif protein 46 | RBM46 | CT68 | 41.6kb | Sense | Chr. 4 - 154781250 - 154828813 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map