Assay Details
| Gene Symbol: LOC100131099 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125931.1 | - | 1 | 751 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Cdk5 and Abl enzyme substrate 2 | CABLES2 |
C20orf150, dJ908M14.2, ik3-2 |
Sense | Chr. 20 - 62388630 - 62407283 on NCBI Build GRCh38 | |
| Downstream | RBBP8 N-terminal like | RBBP8NL | C20orf151 | 4.3kb | Sense | Chr. 20 - 62410237 - 62427573 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100131099
Genomic Map