Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sprouty related EVH1 domain containing 2 | SPRED2 | Spred-2 | Sense | Chr. 2 - 65307425 - 65432599 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC440867 | LOC440867 | 41.8kb | Anti-sense | Chr. 2 - 65436702 - 65459007 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map