Assay Details
| Gene Symbol: FLJ45513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242791.1 | NP_001229720.1 | 2 - 3 | 194 | 77 | |
| GenBank mRNA | AK127421.1 | - | 2 - 3 | 194 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tachykinin 4 (hemokinin) | TAC4 |
EK, HK-1, HK1, PPT-C |
Anti-sense | Chr. 17 - 49838309 - 49848017 on NCBI Build GRCh38 | |
| Downstream | distal-less homeobox 4 | DLX4 |
BP1, DLX7, DLX8, DLX9, OFC15 |
120.4kb | Sense | Chr. 17 - 49968970 - 49974959 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000649.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK127421.1,NM_001242791.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30933621 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10355759 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10355757 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 36359 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3237580 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03255769 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 239400_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008104.hg.1 | ENST00000376609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008104.hg.1 | NM_001242791.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008104.hg.1 | TC17000649.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008218.hg.1 | ENST00000376609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008218.hg.1 | ENST00000376609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008218.hg.1 | ENST00000376609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008218.hg.1 | NM_001242791.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008218.hg.1 | NM_001242791.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008218.hg.1 | NM_001242791.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000503603.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000503603.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000608380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000608380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000608861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000608861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_170685.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_170685.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_170685.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_170685.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_170685.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_170685.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160802.hg.1 | ENST00000376609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160802.hg.1 | NM_001242791.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160802.hg.1 | TC1700008218.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700160802.hg.1 | ENST00000376609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160802.hg.1 | NM_001242791.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008218.hg.1 | ENST00000376609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008218.hg.1 | ENST00000376609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008218.hg.1 | ENST00000376609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008218.hg.1 | NM_001242791.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008218.hg.1 | NM_001242791.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008218.hg.1 | NM_001242791.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000503603.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000503603.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000608380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000608380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000608861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000608861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_170685.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_170685.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_170685.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_170685.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_170685.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_170685.2 | Refseq |
| Human Genome U133 Plus 2.0 Array | 239400_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 239400_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239400_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239400_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239400_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239400_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239400_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239400_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239400_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239400_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239400_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239400_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 239400_at9 | - | - |
| Human Genome U95 Set | 86228_at | - | - |
| Human Genome U95 Set | 86228_PM_at | - | - |
| Human Genome U133 Set | 239400_at | - | - |
| Human Genome U133 Set | 239400_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009684 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009684 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3187848 | NM_001242791.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3187848 | AK127421.1 | human_transcriptome |

FLJ45513
Genomic Map