Assay Details
| Gene Symbol: CD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127925.1 | - | 1 | 2349 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 1 open reading frame 137 | C1orf137 | -58.3kb | Sense | Chr. 1 - 116694112 - 116706603 on NCBI Build GRCh38 | |
| Downstream | prostaglandin F2 receptor inhibitor | PTGFRN |
CD315, CD9P-1, EWI-F, FPRP, SMAP-6 |
142.3kb | Sense | Chr. 1 - 116909923 - 116990358 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CD2
Genomic Map