Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, H/ACA box 71D |
SNORA71D | U71d | -899.0 | Anti-sense | Chr. 20 - 38433862 - 38433999 on NCBI Build GRCh38 |
| Downstream | small nucleolar RNA host gene 11 | SNHG11 |
C20orf198, LINC00101, NCRNA00101 |
9.9kb | Sense | Chr. 20 - 38446654 - 38450921 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map