Assay Details
| Gene Symbol: LOC100128668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128046.1 | - | 1 | 4148 | 69 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DB339412.1 | - | 1 | Chr. 20 - 8252125 - 8252661 [-] on Build GRCh38 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105372520 | LOC105372520 | -219.6kb | Sense | Chr. 20 - 8018870 - 8028623 on NCBI Build GRCh38 | |
| Downstream |
RNA, U105B small nucleolar |
RNU105B |
105B, E1-2 |
578.5kb | Sense | Chr. 20 - 8831185 - 8831393 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100128668
Genomic Map