Assay Details
| Gene Symbol: LOC100130276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128269.1 | - | 1 | 2651 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Dexi homolog | DEXI | MYLE | -121.6kb | Anti-sense | Chr. 16 - 10928891 - 10943007 on NCBI Build GRCh38 |
| Downstream | suppressor of cytokine signaling 1 | SOCS1 |
CIS1, CISH1, JAB, SOCS-1, SSI-1, SSI1, TIP3 |
187.5kb | Anti-sense | Chr. 16 - 11254417 - 11256182 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100130276
Genomic Map