Assay Details
| Gene Symbol: TTC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128840.1 | - | 1 | 3350 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PWWP domain containing 2A | PWWP2A | MST101 | Anti-sense | Chr. 5 - 160043538 - 160120709 on NCBI Build GRCh38 | |
| Downstream | fatty acid binding protein 6 | FABP6 |
I-15P, I-BABP, I-BALB, I-BAP, ILBP, ILBP3, ILLBP |
121.8kb | Sense | Chr. 5 - 160187367 - 160238722 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TTC1
Genomic Map