Assay Details
| Gene Symbol: LOC100128816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY358109.1 | - | 1 | 150 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein arginine methyltransferase 8 | PRMT8 |
HRMT1L3, HRMT1L4 |
-16.3kb | Anti-sense | Chr. 12 - 3381349 - 3593973 on NCBI Build GRCh38 |
| Downstream | calcium release activated channel regulator 2A | CRACR2A | EFCAB4B | 927.0 | Sense | Chr. 12 - 3613366 - 3753216 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591057 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162156 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591060 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591055 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162188 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591042 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162210 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162214 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162227 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162170 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16524962 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162203 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591034 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162172 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162204 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162183 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591013 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16524963 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591066 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162176 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591040 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162189 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591061 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162215 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162190 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591056 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162180 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162186 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162225 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16524969 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591010 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162175 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591025 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591059 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162198 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162153 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591050 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162209 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591036 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162161 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591015 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162164 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162173 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162177 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591047 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591058 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162181 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162219 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591029 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591054 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28591000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162184 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162165 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28590997 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162206 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162167 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16524964 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162163 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08162159 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P931964 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 10904 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 16764 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P931964 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7960363 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7960364 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7960362 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009650.hg.1 | ENST00000624384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009650.hg.1 | ENST00000624384.1 | Ensembl |
| Human Genome U95 Set | 59607_f_at | - | - |
| Human Genome U95 Set | 59607_PM_f_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3266894 | AY358109.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3177615 | AY358109.1 | human_transcriptome |

LOC100128816
Genomic Map