Assay Details
| Gene Symbol: METTL2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY358237.1 | - | 1 | 146 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hypoxia inducible lipid droplet associated | HILPDA |
C7orf68, HIG-2, HIG2 |
-42.3kb | Sense | Chr. 7 - 128455830 - 128458418 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1000 | LINC01000 | 139.7kb | Sense | Chr. 7 - 128641241 - 128660998 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY358237.1 transcripts.
Set Membership: |
|

METTL2B
Genomic Map