Assay Details
| Gene Symbol: CHRFAM7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011522153.1 | XP_011520455.1 | 1 - 2 | 56 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ULK4 pseudogene 1 | ULK4P1 |
D-X, FAM7A1 |
Sense | Chr. 15 - 32406611 - 32435049 on NCBI Build GRCh38 | |
| Downstream | golgin A8 family member R | GOLGA8R | 6.7kb | Sense | Chr. 15 - 30400562 - 30414260 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011522153.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000569278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000569278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000569278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000569278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000569278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000570098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000570098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000570098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000570098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000570098.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000571869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000571869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000571869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000571869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000571869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | ENST00000571869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000397827.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000401522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000562729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000567722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000569278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000569278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000569278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000569278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000569278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000570098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000570098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000570098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000570098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000570098.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000571869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000571869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000571869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000571869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000571869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | ENST00000571869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010841.hg.1 | NM_148911.1 | Refseq |

CHRFAM7A
Genomic Map