Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glutaminyl-peptide cyclotransferase | QPCT |
GCT, QC, sQC |
-298.9kb | Anti-sense | Chr. 2 - 37344610 - 37373322 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 211 | LINC00211 | NCRNA00211 | 153.5kb | Sense | Chr. 2 - 37826247 - 37875863 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map