Assay Details
| Gene Symbol: IL17D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005266421.3 | XP_005266478.1 | 1 | 99 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | intraflagellar transport 88 | IFT88 |
D13S1056E, DAF19, TG737, TTC10, hTg737 |
-11.3kb | Anti-sense | Chr. 13 - 20566446 - 20691437 on NCBI Build GRCh38 |
| Downstream | eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1 | EEF1AKMT1 |
ESP13, N6AMT2 |
25.6kb | Sense | Chr. 13 - 20728931 - 20773958 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005266421.3 transcripts.
Set Membership: |
|

IL17D
Genomic Map