Assay Details
| Gene Symbol: PRDX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF118073.2 | - | 1 | 145 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF118073.2 transcripts.
Set Membership: |
|

PRDX3
Genomic Map