| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017332.hg.1 |
ENST00000414452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017332.hg.1 |
ENST00000414452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017332.hg.1 |
ENST00000414452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017332.hg.1 |
NR_046268.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017332.hg.1 |
NR_046268.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017332.hg.1 |
NR_046268.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100231215.hg.1 |
ENST00000414452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100231215.hg.1 |
NR_046268.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100231215.hg.1 |
ENST00000414452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100231215.hg.1 |
NR_046268.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100231215.hg.1 |
TC0100017332.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100017332.hg.1 |
ENST00000414452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017332.hg.1 |
ENST00000414452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017332.hg.1 |
ENST00000414452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017332.hg.1 |
NR_046268.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017332.hg.1 |
NR_046268.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017332.hg.1 |
NR_046268.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100231211.hg.1 |
ENST00000414452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100231211.hg.1 |
NR_046268.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100231211.hg.1 |
ENST00000414452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100231211.hg.1 |
NR_046268.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100231211.hg.1 |
TC0100017332.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100231213.hg.1 |
ENST00000414452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100231213.hg.1 |
NR_046268.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100231213.hg.1 |
ENST00000414452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100231213.hg.1 |
NR_046268.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100231213.hg.1 |
TC0100017332.hg.1 |
Clariom D TC |