Assay Details
| Gene Symbol: SERPINB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | X98307.1 | - | 1 | 349 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serpin family B member 12 | SERPINB12 | YUKOPIN | -25.8kb | Sense | Chr. 18 - 63519164 - 63569329 on NCBI Build GRCh38 |
| Downstream | serpin family B member 4 | SERPINB4 |
LEUPIN, PI11, SCCA-2, SCCA1, SCCA2 |
41.8kb | Anti-sense | Chr. 18 - 63637259 - 63644319 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SERPINB13
Genomic Map