Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | miR-17-92a-1 cluster host gene | MIR17HG |
C13orf25, FGLDS2, LINC00048, MIHG1, MIRH1, MIRHG1, NCRNA00048, miR-17-92 |
-102.0 | Sense | Chr. 13 - 91347820 - 91354575 on NCBI Build GRCh38 |
| Downstream | glypican 5 | GPC5 | 42.7kb | Sense | Chr. 13 - 91398619 - 92867237 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map