Assay Details
| Gene Symbol: KLHL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK022805.1 | - | 1 | 2944 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interferon alpha 5 | IFNA5 |
IFN-alpha-5, IFN-alphaG, INA5, INFA5, leIF G |
-18.7kb | Sense | Chr. 9 - 21304614 - 21305313 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

KLHL9
Genomic Map