Assay Details
| Gene Symbol: LOC145474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027046.1 | 1 | 1455 | 84 | ||
| GenBank mRNA | AK023718.1 | - | 1 | 1455 | 84 | |
| BQ006454.1 | - | 1 | 84 | |||
| BX102012.1 | - | 1 | 664 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 56B |
SNORD56B | RNU56B | -89.5kb | Sense | Chr. 14 - 71398337 - 71398407 on NCBI Build GRCh38 |
| Downstream | regulator of G-protein signaling 6 | RGS6 | GAP | 442.4kb | Sense | Chr. 14 - 71932100 - 72632478 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000434.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838028 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838060 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378515 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838032 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378510 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378537 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378514 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838050 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378533 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378485 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378486 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838043 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838044 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838061 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838049 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378522 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378491 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838041 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838053 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378502 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838033 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838031 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838030 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838068 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378507 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378531 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838071 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838070 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838073 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838048 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378493 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838054 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378535 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838038 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838056 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838057 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378539 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838042 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838080 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378496 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378521 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29838052 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09378525 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 814817 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3327697 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4958 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4959 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 13406 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3327697 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3327697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16714737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378527 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378493 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838049 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838073 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378537 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838062 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838054 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838052 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838060 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378515 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378496 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838047 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378502 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378531 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378535 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838029 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838079 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378492 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838038 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378539 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838050 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378525 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838050 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378537 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16714737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378531 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838038 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838060 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378492 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378496 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378493 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378527 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838047 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838073 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378539 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838054 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838049 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378515 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838062 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838052 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378535 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838029 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378502 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838079 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09378525 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29838081 | - | - |
| Agilent Human 2 cDNA Microarray (G4101A) | 3386677 | - | - |
| Agilent Human 2 cDNA Microarray (G4101A) | 3386677 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P393377 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378570 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378562 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378554 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16714743 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378508 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378515 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378507 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378551 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378580 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378503 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378485 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378549 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378547 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378501 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378577 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16714738 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378567 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378489 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378522 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378492 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378506 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378517 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378494 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378526 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378487 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378553 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378529 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378532 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378534 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378499 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378524 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378583 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378530 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378541 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378560 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378575 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378558 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378573 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378545 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378496 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378519 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378555 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378511 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378582 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09378538 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 230505_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005229.hg.1 | NR_027046.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005229.hg.1 | TC14000434.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400153328.hg.1 | NR_027046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153328.hg.1 | NR_027046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153328.hg.1 | TC1400007595.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400153326.hg.1 | NR_027046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153326.hg.1 | NR_027046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153326.hg.1 | TC1400007595.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400153327.hg.1 | NR_027046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153327.hg.1 | NR_027046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153327.hg.1 | TC1400007595.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 230505_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 230505_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230505_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230505_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230505_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230505_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230505_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230505_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230505_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230505_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230505_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230505_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230505_at7 | - | - |
| Human Genome U95 Set | 88386_PM_at | - | - |
| Human Genome U95 Set | 88386_at | - | - |
| Human Genome U133 Set | 230505_at | - | - |
| Human Genome U133 Set | 230505_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051572 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051572 | - | - |

LOC145474
Genomic Map