Assay Details
| Gene Symbol: USP34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK024341.1 | - | 1 | 1251 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast) |
AHSA2 | Hch1 | -35.1kb | Anti-sense | Chr. 2 - 61177418 - 61191210 on NCBI Build GRCh38 |
| Downstream |
small nucleolar RNA, H/ACA box 70B |
SNORA70B | 191.6kb | Sense | Chr. 2 - 61417244 - 61417377 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

USP34
Genomic Map