Assay Details
| Gene Symbol: PPP1R16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK026900.1 | - | 1 | 1253 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ARP5 actin-related protein 5 homolog | ACTR5 |
Arp5, INO80M |
-66.9kb | Sense | Chr. 20 - 38748454 - 38772446 on NCBI Build GRCh38 |
| Downstream | family with sequence similarity 83 member D | FAM83D |
C20orf129, CHICA, dJ616B8.3 |
83.7kb | Sense | Chr. 20 - 38926312 - 38953060 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PPP1R16B
Genomic Map