Assay Details
| Gene Symbol: MESP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL357535.1 | - | 1 | 1154 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WD repeat domain 93 | WDR93 | -4.9kb | Anti-sense | Chr. 15 - 89690074 - 89744390 on NCBI Build GRCh38 | |
| Downstream | mesoderm posterior bHLH transcription factor 2 | MESP2 |
SCDO2, bHLHc6 |
26.2kb | Anti-sense | Chr. 15 - 89776358 - 89778754 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MESP1
Genomic Map