Assay Details
| Gene Symbol: LOC100294362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029376.1 | 2 | 2766 | 66 | ||
| GenBank mRNA | AF318359.1 | - | 1 | 1048 | 66 | |
| AK125341.1 | - | 2 | 2717 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ring finger protein 213 | RNF213 |
ALO17, C17orf27, KIAA1618, MYMY2, MYSTR, NET57 |
Anti-sense | Chr. 17 - 80260820 - 80398786 on NCBI Build GRCh38 | |
| Downstream | endonuclease V | ENDOV | 61.3kb | Anti-sense | Chr. 17 - 80415117 - 80438086 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001935.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 366035 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 11475 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 422251 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 442617 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 888362 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 629872 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P372074 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 27402 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074598 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488274 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074597 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074616 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488285 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074594 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074595 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074622 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488262 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074599 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074603 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074605 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488276 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074598 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488274 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074603 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074595 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488262 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074599 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074605 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488285 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074594 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074622 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488276 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074616 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10488267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31074597 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P372074 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03298315 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20742638 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P372074 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03298315 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20742638 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P20742638 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03298315 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8010464 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8010461 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8019072 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8019073 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8010462 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8010463 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8019071 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1569078_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000572151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000572151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000573394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000573394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000573394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | ENST00000613190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011974.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026379.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026379.hg.1 | TC17001935.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026380.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026380.hg.1 | TC17001935.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700197859.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197859.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197859.hg.1 | ENST00000613190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197859.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197859.hg.1 | TC1700011974.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197859.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197859.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197859.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197859.hg.1 | ENST00000613190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197865.hg.1 | ENST00000573394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197865.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197865.hg.1 | TC1700011974.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197865.hg.1 | ENST00000573394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197865.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197860.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197860.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197860.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197860.hg.1 | TC1700011974.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197860.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197860.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197860.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197864.hg.1 | ENST00000572151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197864.hg.1 | ENST00000573394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197864.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197864.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197864.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197864.hg.1 | TC1700011974.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197864.hg.1 | ENST00000572151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197864.hg.1 | ENST00000573394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197864.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197864.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197864.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197861.hg.1 | ENST00000572151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197861.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197861.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197861.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197861.hg.1 | TC1700011974.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197861.hg.1 | ENST00000572151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197861.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197861.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197861.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197862.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197862.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197862.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700197862.hg.1 | TC1700011974.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700197862.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197862.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700197862.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000572151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000572151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000573394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000573394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000573394.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000575034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | ENST00000613190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | NR_029376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011974.hg.1 | NR_029376.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1569078_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1569078_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569078_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569078_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569078_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569078_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569078_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569078_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569078_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569078_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569078_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569078_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569078_at3 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008869 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008868 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008869 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008868 | - | - |

LOC100294362
Genomic Map